accessibility menu, dialog, popup

UserName

CAS RN 13678-68-7

H3COSO

CAS RN 13678-68-7

IUPAC Name: 1-{[(furan-2-yl)methyl]sulfanyl}ethan-1-one
Synonyms: 1-[(furan-2-ylmethyl)sulfanyl]ethanone
Molecular Weight (g/mol): 156.2
Molecular Formula: C7H8O2S
InChi Key: LQOUTUIIYXYBQW-UHFFFAOYSA-N
SMILES: CC(=O)SCC1=CC=CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results