missing translation for 'onlineSavingsMsg'
Learn More

S-(-)-Etomidate, TRC

Product Code. 30373802
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373802 100 mg 100mg
30369203 10 mg -
30386477 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30373802 Supplier Toronto Research Chemicals Supplier No. TRCE933305100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56649-47-9
Molecular Formula C14H16N2O2
Molecular Weight (g/mol) 244.29
Synonym 1-[(1S)-1-Phenylethyl]-1H-Imidazole-5-carboxylic Acid Ethyl Ester, (S)-1-(1-Phenylethyl)-,1H-imidazole-5-carboxylic Acid Ethyl Ester, S(-)-Etomidate, (S)-Etomidate, l-Etomidate
SMILES CCOC(=O)c1cncn1[C@@H](C)c2ccccc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H16N2O2/c1-3-18-14(17)13-9-15-10-16(13)11(2)12-7-5-4-6-8-12/h4-11H,3H2,1-2H3/t11-/m0/s1
Formula Weight 244.1212
Chemical Name or Material S-(-)-Etomidate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.