CAS RN 56649-47-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
S-(-)-Etomidate, TRC
CAS: 56649-47-9 Molecular Formula: C14H16N2O2 Molecular Weight (g/mol): 244.29 Synonym: 1-[(1S)-1-Phenylethyl]-1H-Imidazole-5-carboxylic Acid Ethyl Ester,(S)-1-(1-Phenylethyl)-,1H-imidazole-5-carboxylic Acid Ethyl Ester,S(-)-Etomidate,(S)-Etomidate,l-Etomidate SMILES: CCOC(=O)c1cncn1[C@@H](C)c2ccccc2