missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (S)-Crizotinib, >98%

Description
Potent MTH1 inhibitor (IC50 = 72 nM). Induces DNA damage and disrupts nucleotide pool homeostasis in cancer cells. Also attenuates colony formation of KRAS-mutated PANC1 cells in vitro. Suppresses tumor growth ∽50% in a colon cancer carcinoma xenograft model.
Specifications
Specifications
| Quantity | 10 mg |
| Solubility | Soluble to 50mM in DMSO and to 20mM in 1eq. HCl with gentle warming |
| Formula Weight | 450.34 |
| Purity | >98% |
| Molecular Weight (g/mol) | 450.339 |
| Chemical Name or Material | 3-[(1S)-1-(2,6-Dichloro-3-fluorophenyl)ethoxy]-5-[1-(4-piperidinyl)-1H-pyrazol-4-yl]-2-pyridinamine |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?