accessibility menu, dialog, popup

UserName

CAS RN 1374356-45-2

CH3ClFClONNNNHNH2

CAS RN 1374356-45-2

IUPAC Name: 3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]-5-[1-(piperidin-4-yl)-1H-pyrazol-4-yl]pyridin-2-amine
Synonyms: crizotinib
Molecular Weight (g/mol): 450.34
Molecular Formula: C21H22Cl2FN5O
InChi Key: KTEIFNKAUNYNJU-UHFFFAOYNA-N
SMILES: CC(OC1=C(N)N=CC(=C1)C1=CN(N=C1)C1CCNCC1)C1=C(Cl)C=CC(F)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results