missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Alpha-Lipoic Acid, TRC

Product Code. 17970985
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17970985 25 mg 25mg
17964355 100 mg 100mg
17960885 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970985 Supplier Toronto Research Chemicals Supplier No. TRCL46873025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1077-27-6
Molecular Formula C8H14O2S2
Molecular Weight (g/mol) 206.33
Synonym (3S)-1,2-Dithiolane-3-pentanoic Acid, (-)-Thioctic Acid, (S)-(-)-Lipoic Acid, (S)-Lipoic Acid, (S)-Thioctic Acid, l-Thioctic Acid;
SMILES C1CSS[C@H]1CCCCC(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H14O2S2/c9-8(10)4-2-1-3-7-5-6-11-12-7/h7H,1-6H2,(H,9,10)/t7-/m0/s1
Percent Purity >95%
Chemical Name or Material (S)-(-)-alpha-Lipoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.