CAS RN 1077-27-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-(-)-Alpha-Lipoic Acid, TRC
CAS: 1077-27-6 Molecular Formula: C8H14O2S2 Molecular Weight (g/mol): 206.33 Synonym: (3S)-1,2-Dithiolane-3-pentanoic Acid,(-)-Thioctic Acid,(S)-(-)-Lipoic Acid,(S)-Lipoic Acid,(S)-Thioctic Acid,l-Thioctic Acid; SMILES: C1CSS[C@H]1CCCCC(=O)O