missing translation for 'onlineSavingsMsg'
Learn More

(S)-6-Hydroxy Nicotine, TRC

Product Code. 30397002
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397002 5 mg -
30397428 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397002 Supplier Toronto Research Chemicals Supplier No. TRCH9485555MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10516-09-3
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.23
Synonym 5-[(2S)-1-Methyl-2-pyrrolidinyl]-2(1H)-pyridinone, (-)-5-(1-Methyl-2-pyrrolidinyl) -2-pyridinol, 6-Hydroxynicotine, L-6-Hydroxynicotine, 6-Hydroxynicotine
SMILES Ic1ccc(cc1)c2n(nc(C(=O)NN3CCCCC3)c2C)c4c(Cl)cc(Cl)cc4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H14N2O/c1-12-6-2-3-9(12)8-4-5-10(13)11-7-8/h4-5,7,9H,2-3,6H2,1H3,(H,11,13)/t9-/m0/s1
Chemical Name or Material (S)-6-Hydroxy Nicotine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.