CAS RN 10516-09-3
CAS RN 10516-09-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-6-Hydroxy Nicotine, TRC
CAS: 10516-09-3 Molecular Formula: C10H14N2O Molecular Weight (g/mol): 178.23 Synonym: 5-[(2S)-1-Methyl-2-pyrrolidinyl]-2(1H)-pyridinone,(-)-5-(1-Methyl-2-pyrrolidinyl) -2-pyridinol,6-Hydroxynicotine,L-6-Hydroxynicotine,6-Hydroxynicotine SMILES: Ic1ccc(cc1)c2n(nc(C(=O)NN3CCCCC3)c2C)c4c(Cl)cc(Cl)cc4