missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-Dimethylaminopyrrolidine dihydrochloride, 97%, Thermo Scientific™

Product Code. 15433977
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433977 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15433977 Supplier Thermo Scientific Alfa Aesar Supplier No. H30661.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144043-20-9
Molecular Formula C6H16Cl2N2
Molecular Weight (g/mol) 187.11
MDL Number MFCD08561117
InChI Key LCPKWRSLMCUOOZ-UHFFFAOYNA-N
Synonym s-3-dimethylaminopyrrolidine dihydrochloride, s-3-dimethylaminopyrrolidine 2hcl, s-n,n-dimethylpyrrolidin-3-amine dihydrochloride, 3s-n,n-dimethylpyrrolidin-3-amine dihydrochloride, s---3-dimethylaminopyrrolidine dihydrochloride, s---3-dimethylaminopyrroli, 3s-n,n-dimethylpyrrolidin-3-amine hydrochloride, s-n,n-dimethyl-3-pyrrolidinamine.2hcl, 3s-3-dimethylamino pyrrolidine dihydrochloride, s-+-n,n-dimethyl-3-pyrrolidinamine dihydrochloride
PubChem CID 53482061
IUPAC Name N,N-dimethylpyrrolidin-3-amine dihydrochloride
SMILES Cl.Cl.CN(C)C1CCNC1

Specifications

Melting Point 176°C to 181°C
Quantity 250 mg
Sensitivity Hygroscopic
Optical Rotation −3.7° (c=1 in Water)
Formula Weight 187.11
Percent Purity 97%
Chemical Name or Material (S)-(-)-3-Dimethylaminopyrrolidine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.