accessibility menu, dialog, popup

UserName

CAS RN 144043-20-9

H3CNCH3HNHClHCl

CAS RN 144043-20-9

IUPAC Name: N,N-dimethylpyrrolidin-3-amine dihydrochloride
Synonyms: N,N-dimethylpyrrolidin-3-amine dihydrochloride
Molecular Weight (g/mol): 187.11
Molecular Formula: C6H16Cl2N2
InChi Key: LCPKWRSLMCUOOZ-UHFFFAOYNA-N
SMILES: Cl.Cl.CN(C)C1CCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results