missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-1-benzylpiperidine, 97%

Product Code. 15459507
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459507 5 g 5g
15449507 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459507 Supplier Thermo Scientific Alfa Aesar Supplier No. H52772.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 168466-85-1 | C12H18N2 | 190.29 g/mol

(S)-3-Amino-1-benzylpiperidine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Amino-1-benzylpiperidine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Freely soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168466-85-1
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD03093375
InChI Key HARWNWOLWMTQCC-LBPRGKRZSA-N
Synonym s-1-benzylpiperidin-3-amine, s-3-amino-1-benzylpiperidine, 3s-1-benzylpiperidin-3-amine, s-1-benzyl-3-aminopiperidine, s-3-amino-1-benzyl-piperidine, 3-piperidinamine, 1-phenylmethyl-, 3s, s-n-benzyl-3-amino-piperidine, s-1-benzyl-piperidine-3-ylamine, s-3-amino-n-benzylpiperidine, pubchem11490
PubChem CID 854131
IUPAC Name (3S)-1-benzylpiperidin-3-amine
SMILES C1CC(CN(C1)CC2=CC=CC=C2)N

Specifications

Quantity 5 g
Solubility Information Freely soluble in ethanol.
Formula Weight 190.29
Percent Purity 97%
Chemical Name or Material (S)-3-Amino-1-benzylpiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.