accessibility menu, dialog, popup

UserName

CAS RN 168466-85-1

NH2N

CAS RN 168466-85-1

IUPAC Name: 1-benzylpiperidin-3-amine
Synonyms: 1-benzylpiperidin-3-amine
Molecular Weight (g/mol): 190.29
Molecular Formula: C12H18N2
InChi Key: HARWNWOLWMTQCC-UHFFFAOYNA-N
SMILES: NC1CCCN(CC2=CC=CC=C2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results