missing translation for 'onlineSavingsMsg'
Learn More

(S)-3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dimethoxybenzamide, TRC

Product Code. 30383629
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383629 1 g
30379556 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383629 Supplier Toronto Research Chemicals Supplier No. TRCD4363801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84225-94-5
Molecular Formula C16H22Cl2N2O3
Molecular Weight (g/mol) 361.26
Synonym 3,5-Dichloro-N-[[(2S)-1-ethyl-2-pyrrolidinyl]methyl]-2,6-dimethoxybenzamide
SMILES CCN1CCC[C@@H]1CNC(=O)c2c(OC)c(Cl)cc(Cl)c2OC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H22Cl2N2O3/c1-4-20-7-5-6-10(20)9-19-16(21)13-14(22-2)11(17)8-12(18)15(13)23-3/h8,10H,4-7,9H2,1-3H3,(H,19,21)
Formula Weight 360.1007
Chemical Name or Material (S)-3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dimethoxybenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.