CAS RN 84225-94-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-3,5-Dichloro-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2,6-dimethoxybenzamide, TRC
CAS: 84225-94-5 Molecular Formula: C16H22Cl2N2O3 Molecular Weight (g/mol): 361.26 Synonym: 3,5-Dichloro-N-[[(2S)-1-ethyl-2-pyrrolidinyl]methyl]-2,6-dimethoxybenzamide SMILES: CCN1CCC[C@@H]1CNC(=O)c2c(OC)c(Cl)cc(Cl)c2OC