missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Hydroxybutanoic Acid, TRC

Product Code. 30399271
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399271 5 g -
30402904 1 g 1g
30393007 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399271 Supplier Toronto Research Chemicals Supplier No. TRCH9563285G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3347-90-8
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.1
Synonym (2S)-2-Hydroxybutanoic Acid, (+)-2-Hydroxy-n-butyric Acid, (+)-2-Hydroxybutanoic Acid, (+)-2-Hydroxybutyric Acid, (+)-alpha-Hydroxybutyric Acid, (2S)-2-Hydroxybutanoic Acid, (S)-(+)-2-Hydroxybutanoic Acid, (S)-2-Hydroxybutanoic Acid, (S)-2-Hydroxybutyric Acid, L-2-Hydroxybutyric Acid, L-alpha-Hydroxybutyric Acid
IUPAC Name (2S)-2-hydroxybutanoic acid
SMILES CC[C@H](O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H8O3/c1-2-3(5)4(6)7/h3,5H,2H2,1H3,(H,6,7)/t3-/m0/s1
Formula Weight 104.0473
Chemical Name or Material (S)-2-Hydroxybutanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.