CAS RN 3347-90-8
IUPAC Name:
2-hydroxybutanoic acid
Synonyms:
α-hydroxybutyric acid (+/-)α-hydoxy butyric acid
Molecular Weight (g/mol):
104.11
Molecular Formula:
C4H8O3
InChi Key:
AFENDNXGAFYKQO-UHFFFAOYNA-N
SMILES:
CCC(O)C(O)=O
1
–
1
of
1
results
1
(S)-2-Hydroxybutanoic Acid, TRC
CAS: 3347-90-8 Molecular Formula: C4H8O3 Molecular Weight (g/mol): 104.1 Synonym: (2S)-2-Hydroxybutanoic Acid,(+)-2-Hydroxy-n-butyric Acid,(+)-2-Hydroxybutanoic Acid,(+)-2-Hydroxybutyric Acid,(+)-alpha-Hydroxybutyric Acid,(2S)-2-Hydroxybutanoic Acid,(S)-(+)-2-Hydroxybutanoic Acid,(S)-2-Hydroxybutanoic Acid,(S)-2-Hydroxybutyric Acid,L-2-Hydroxybutyric Acid,L-alpha-Hydroxybutyric Acid IUPAC Name: (2S)-2-hydroxybutanoic acid SMILES: CC[C@H](O)C(=O)O