missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Heptanol, 98%

Product Code. 11337778
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337778 250 mg 250mg
11347778 1 g 1g
15451337 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11337778 Supplier Thermo Scientific Alfa Aesar Supplier No. 033795.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6033-23-4 | C7H16O | 116.204 g/mol

(S)-(+)-2-Heptanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6033-23-4
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00065003
InChI Key CETWDUZRCINIHU-ZETCQYMHSA-N
Synonym s-+-2-heptanol, 2s-heptan-2-ol, 2-heptanol, s, unii-7xq9f7kypw, heptan-2s-ol, s-2-heptanol, s-heptan-2-ol, 7xq9f7kypw, d-2-heptanol
PubChem CID 2724897
ChEBI CHEBI:87378
IUPAC Name (2S)-heptan-2-ol
SMILES CCCCCC(C)O

Specifications

Density 0.815 g/mL
Boiling Point 160°C to 162°C
Flash Point 59°C (138°F)
Refractive Index 1.421
Quantity 250 mg
UN Number UN1987
Merck Index 14,4662
Solubility Information Soluble in water(3569mg/L at 25°C).
Formula Weight 116.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-Heptanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.