accessibility menu, dialog, popup

UserName

H3CCH3OH

CAS RN 6033-23-4

IUPAC Name: heptan-2-ol
Synonyms: 2-heptanol
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: CETWDUZRCINIHU-UHFFFAOYNA-N
SMILES: CCCCCC(C)O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)