missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Heptanol, 98%

Product Code. 11347778
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11347778

Brand: Thermo Scientific Alfa Aesar 033795.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6033-23-4 | C7H16O | 116.204 g/mol

(S)-(+)-2-Heptanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6033-23-4
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00065003
InChI Key CETWDUZRCINIHU-ZETCQYMHSA-N
Synonym s-+-2-heptanol, 2s-heptan-2-ol, 2-heptanol, s, unii-7xq9f7kypw, heptan-2s-ol, s-2-heptanol, s-heptan-2-ol, 7xq9f7kypw, d-2-heptanol
PubChem CID 2724897
ChEBI CHEBI:87378
IUPAC Name (2S)-heptan-2-ol
SMILES CCCCCC(C)O

Specifications

Density 0.815 g/mL
Boiling Point 160°C to 162°C
Flash Point 59°C (138°F)
Refractive Index 1.421
Quantity 1 g
UN Number UN1987
Merck Index 14,4662
Solubility Information Soluble in water(3569mg/L at 25°C).
Formula Weight 116.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-Heptanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.