missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-1-BOC-3-(Hydroxymethyl)piperidine, 97%, Thermo Scientific™
£34.69 - £122.01
Chemical Identifiers
CAS | 140695-84-7 |
---|---|
Molecular Formula | C11H21NO3 |
Molecular Weight (g/mol) | 215.29 |
InChI Key | OJCLHERKFHHUTB-VIFPVBQESA-N |
Synonym | s-1-boc-3-hydroxymethyl piperidine, s-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, s-n-boc-3-piperidinemethanol, tert-butyl 3s-3-hydroxymethyl piperidine-1-carboxylate, s-1-n-boc-3-hydroxymethyl-piperidine, s-1-boc-3-hyroxymethyl piperidine, s-1-n-boc-3-hydroxymethyl piperidine, s-n-boc-3-hyroxymethyl piperidine, s-1-boc-3-hydroxymethylpiperidine |
PubChem CID | 1514448 |
IUPAC Name | tert-butyl (3S)-3-(hydroxymethyl)piperidine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCCC(C1)CO |
Description
Chemical Identifiers
140695-84-7 | |
215.29 | |
s-1-boc-3-hydroxymethyl piperidine, s-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, s-n-boc-3-piperidinemethanol, tert-butyl 3s-3-hydroxymethyl piperidine-1-carboxylate, s-1-n-boc-3-hydroxymethyl-piperidine, s-1-boc-3-hyroxymethyl piperidine, s-1-n-boc-3-hydroxymethyl piperidine, s-n-boc-3-hyroxymethyl piperidine, s-1-boc-3-hydroxymethylpiperidine | |
tert-butyl (3S)-3-(hydroxymethyl)piperidine-1-carboxylate |
C11H21NO3 | |
OJCLHERKFHHUTB-VIFPVBQESA-N | |
1514448 | |
CC(C)(C)OC(=O)N1CCCC(C1)CO |
Specifications
140695-84-7 | |
100.0 | |
White | |
96% min. (GC) | |
15 | |
OJCLHERKFHHUTB-VIFPVBQESA-N | |
tert-butyl (3S)-3-(hydroxymethyl)piperidine-1-carboxylate | |
1514448 | |
97% | |
(S)-1-BOC-3-(Hydroxymethyl)piperidine |
96.0 | |
92.3°C to 94.2°C | |
Authentic | |
C11H21NO3 | |
s-1-boc-3-hydroxymethyl piperidine, s-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, s-n-boc-3-piperidinemethanol, tert-butyl 3s-3-hydroxymethyl piperidine-1-carboxylate, s-1-n-boc-3-hydroxymethyl-piperidine, s-1-boc-3-hyroxymethyl piperidine, s-1-n-boc-3-hydroxymethyl piperidine, s-n-boc-3-hyroxymethyl piperidine, s-1-boc-3-hydroxymethylpiperidine | |
CC(C)(C)OC(=O)N1CCCC(C1)CO | |
215.29 | |
215.29 | |
Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- (S)-1-BOC-3-(Hydroxymethyl)piperidine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only