missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-BOC-3-(Hydroxymethyl)piperidine, 97%, Thermo Scientific™

Product Code. 10119799 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10119799 5 g Glass bottle 5g
10068049 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10119799 Supplier Thermo Scientific Acros Supplier No. 437480050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 140695-84-7
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.29
InChI Key OJCLHERKFHHUTB-VIFPVBQESA-N
Synonym s-1-boc-3-hydroxymethyl piperidine, s-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, s-n-boc-3-piperidinemethanol, tert-butyl 3s-3-hydroxymethyl piperidine-1-carboxylate, s-1-n-boc-3-hydroxymethyl-piperidine, s-1-boc-3-hyroxymethyl piperidine, s-1-n-boc-3-hydroxymethyl piperidine, s-n-boc-3-hyroxymethyl piperidine, s-1-boc-3-hydroxymethylpiperidine
PubChem CID 1514448
IUPAC Name tert-butyl (3S)-3-(hydroxymethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 92.3°C to 94.2°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation 15
Formula Weight 215.29
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (S)-1-BOC-3-(Hydroxymethyl)piperidine
Show More Show Less
compliance-icons
Product Identifier
  • (S)-1-BOC-3-(Hydroxymethyl)piperidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.