Learn More
RUVBL2 Mouse, anti-Human, Unconjugated, Clone: 1A1A4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RUVBL2 |
|---|---|
| Clone | 1A1A4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17251516
|
Proteintech
67851-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17241516
|
Proteintech
67851-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RUVBL2 belongs to the conserved ATPases associated with various cellular activities (AAA+) protein subfamily, which is characterized by the presence of conserved Walker A and B motifs that are involved in ATP binding and hydrolysis. The AAA+ (ATPases associated with diverse cellular activities) ATPases, RUVBL1 and RUVBL2, were originally isolated as components of transcriptional complexes and shown to function in a number of different cellular processes, including transcriptional regulation, chromatin remodelling and DNA damage responses. (PMID: 31572066, PMID: 31138842)
Specifications
| RUVBL2 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| Primary |
| 1A1A4 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| 10856 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.