Learn More
RUVBL2 Mouse, anti-Human, Unconjugated, Clone: 1A1A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67851-1-IG-150UL
Description
RUVBL2 belongs to the conserved ATPases associated with various cellular activities (AAA+) protein subfamily, which is characterized by the presence of conserved Walker A and B motifs that are involved in ATP binding and hydrolysis. The AAA+ (ATPases associated with diverse cellular activities) ATPases, RUVBL1 and RUVBL2, were originally isolated as components of transcriptional complexes and shown to function in a number of different cellular processes, including transcriptional regulation, chromatin remodelling and DNA damage responses. (PMID: 31572066, PMID: 31138842)
Specifications
| RUVBL2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1A1A4 | |
| Unconjugated | |
| 150 μL | |
| 10856 | |
| Liquid |