missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ruthenium Red tetrahydrate, pure

Product Code. 10400091 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10400091 1 g Glass bottle 1g
10773255 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10400091 Supplier Thermo Scientific Chemicals Supplier No. 191730010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12790-48-6 | Cl6H36N14O6Ru3-14 | 844.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12790-48-6
Molecular Formula Cl6H36N14O6Ru3-14
Molecular Weight (g/mol) 844.29
MDL Number MFCD00167176
InChI Key IMVKVBRYNZXYAP-UHFFFAOYSA-H
PubChem CID 108195
IUPAC Name azanide;oxygen(2-);ruthenium;ruthenium(5+);hexachloride;tetrahydrate
SMILES [NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].O.O.O.O.[O-2].[O-2].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru].[Ru+5].[Ru+5]

Specifications

Linear Formula [(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6·4H2O
Merck Index 10, 8161
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ammonia
Lambda Max 530 to 536nm
Formula Weight 858.35
Percent Purity 34.6 to 36.1% (ruthenium)
Grade Pure
Molar Absorption Coefficient 8000L/(mole.cm) (lambda min., 0.02g/L H2O)
Packaging Glass bottle
Color Purple/Red
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Ruthenium red tetrahydrate, Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.