CAS RN 12790-48-6
IUPAC Name:
bis(λ⁵-ruthenium(5+)) tetrahydrate ruthenium tetradecaazanide hexachloride dioxidandiide
Synonyms:
bis(λ⁵-ruthenium(5+)) tetrahydrate ruthenium tetradecaazanide hexachloride dioxidandiide
Molecular Weight (g/mol):
844.3
Molecular Formula:
Cl6H36N14O6Ru3
InChi Key:
IMVKVBRYNZXYAP-UHFFFAOYSA-H
SMILES:
[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].O.O.O.O.[O--].[O--].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru].[Ru+5].[Ru+5]
1
–
1
of
1
results
1
Thermo Scientific Chemicals Ruthenium Red tetrahydrate, pure
CAS: 12790-48-6 Molecular Formula: Cl6H36N14O6Ru3-14 Molecular Weight (g/mol): 844.29 MDL Number: MFCD00167176 InChI Key: IMVKVBRYNZXYAP-UHFFFAOYSA-H PubChem CID: 108195 IUPAC Name: azanide;oxygen(2-);ruthenium;ruthenium(5+);hexachloride;tetrahydrate SMILES: [NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].O.O.O.O.[O-2].[O-2].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru].[Ru+5].[Ru+5]