missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX1/2/3 Antibody (SR1906), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | RUNX1/2/3 |
|---|---|
| Clone | SR1906 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18628374
|
Novus Biologicals
NBP3-21666-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611025
|
Novus Biologicals
NBP3-21666-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RUNX1/2/3 Monoclonal antibody specifically detects RUNX1/2/3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
| RUNX1/2/3 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Acute myeloid leukemia 1, 2, 3 protein, AML1, 2, 3, AMLCR1, CBFA1, 2, 3, CBF-alpha-1, 2, 3, CCD, Core-binding factor subunit alpha-1, 2, 3, EVI-1, Oncogene AML-1, 2, 3, PEA2-alpha A, B, C, PEBP2-alpha A, B, C, Polyomavirus enhancer-binding protein 2 alpha A, B, C subunit, Runt-related transcription factor 1, 2, 3, RUNX family transcription factor 1, 2, 3, SL3/AKV core-binding factor alpha A, B, C subunit, SL3-3 enhancer factor 1 alpha A, B, C subunit | |
| A synthesized peptide derived from human RUNX1/2/3 (Uniprot #: Q01196, Q13761, Q13950) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1906 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Neuroscience, Protein Phosphatase, Stem Cells | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 861 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title