missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX1/2/3 Antibody (SR1906), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21666-100ul
This item is not returnable.
View return policy
Description
RUNX1/2/3 Monoclonal antibody specifically detects RUNX1/2/3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| RUNX1/2/3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR1906 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Acute myeloid leukemia 1, 2, 3 protein, AML1, 2, 3, AMLCR1, CBFA1, 2, 3, CBF-alpha-1, 2, 3, CCD, Core-binding factor subunit alpha-1, 2, 3, EVI-1, Oncogene AML-1, 2, 3, PEA2-alpha A, B, C, PEBP2-alpha A, B, C, Polyomavirus enhancer-binding protein 2 alpha A, B, C subunit, Runt-related transcription factor 1, 2, 3, RUNX family transcription factor 1, 2, 3, SL3/AKV core-binding factor alpha A, B, C subunit, SL3-3 enhancer factor 1 alpha A, B, C subunit | |
| A synthesized peptide derived from human RUNX1/2/3 (Uniprot #: Q01196, Q13761, Q13950) | |
| 100 μg | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Neuroscience, Protein Phosphatase, Stem Cells | |
| 861 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction