missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Twistit Stoppers, Rubber
Use these versatile Rubber Twistit Stoppers as solid, one-hole, two-hole, or three-hole stoppers.
Brand: Cole-Parmer™ 62992-30
Code : A0
Additional Details : Weight : 0.42300kg
Description
- Simply twist off a bottom seal to open a hole.
- The soft resilient natural rubber stays pliable over a long period and withstands laboratory conditions without hardening.
- Stoppers retain resiliency even after repeated autoclaving.
- Each stopper has its size in raised figures on top.
- Available in black or white rubber.
- Autoclavable.
Specifications
White | |
25 mm | |
37 mm | |
12 Pk. | |
Autoclavable |
Twist it Stopper | |
9 | |
45 mm | |
Rubber |