missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSPH10B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifica
| Antigen | RSPH10B |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Descrizione
RSPH10B Polyclonal antibody specifically detects RSPH10B in Human samples. It is validated for Western BlotSpecifica
| RSPH10B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 222967 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC50833, radial spoke head 10 homolog B, radial spoke head 10 homolog B (Chlamydomonas), radial spoke head 10 homolog B2, RSPH10B2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RSPH10B (NP_775836.3). MVKEKKKADKKGEKSARSPSSLSDNLDFSKQDGNTTRQEMSPAGVPLLGMQLNEVKPKKDRQNVQQNEDATQYEESILTKLIVESYEGEKVRGLYEGEGFAAFQGGCTYRGMFSEGLMHGQGTYIWADGLKYEGDFVKNVPMNHGVYTWPDGSMYEGEVVNGMRNGFGMFKCSTQPVSYIGHWCNGKRHGKGSIYYNQEGTCWYEGDWVQNIKKGWGIRCYKSGNIYEGQWEDNMRHGEGRMRWLTTNEEYTGRWERGIQNGFGTHTWFLKRIRSSQYPLRNEYIGEFVNGYRHGRGKFY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto