missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSPH10B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94037-0.1ml
This item is not returnable.
View return policy
Description
RSPH10B Polyclonal antibody specifically detects RSPH10B in Human samples. It is validated for Western Blot
Specifications
| RSPH10B | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC50833, radial spoke head 10 homolog B, radial spoke head 10 homolog B (Chlamydomonas), radial spoke head 10 homolog B2, RSPH10B2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RSPH10B (NP_775836.3). MVKEKKKADKKGEKSARSPSSLSDNLDFSKQDGNTTRQEMSPAGVPLLGMQLNEVKPKKDRQNVQQNEDATQYEESILTKLIVESYEGEKVRGLYEGEGFAAFQGGCTYRGMFSEGLMHGQGTYIWADGLKYEGDFVKNVPMNHGVYTWPDGSMYEGEVVNGMRNGFGMFKCSTQPVSYIGHWCNGKRHGKGSIYYNQEGTCWYEGDWVQNIKKGWGIRCYKSGNIYEGQWEDNMRHGEGRMRWLTTNEEYTGRWERGIQNGFGTHTWFLKRIRSSQYPLRNEYIGEFVNGYRHGRGKFY | |
| 0.1 mL | |
| Cell Biology | |
| 222967 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction