missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSBN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57724
This item is not returnable.
View return policy
Description
RSBN1 Polyclonal specifically detects RSBN1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| RSBN1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DKFZp781E21150, FLJ11220, ROSBIN, round spermatid basic protein 1, RP11-324J2.1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54665 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, KnockDown | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, KnockDown Validated | |
| Q5VWQ0 | |
| RSBN1 | |
| Synthetic peptides corresponding to RSBN1 (round spermatid basic protein 1) The peptide sequence was selected from the N terminal of RSBN1)(50ug). Peptide sequence GGAVGPFKCVFVGEMAAQVGAVRVVRAVAAQEEPDKEGKEKPHAGVSPRG. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Bovine: 85%. | |
| Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction