missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (RS)-MCPG
GREENER_CHOICE

Product Code. 18083852
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18083852 10 mg 10mg
30001080 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18083852 Supplier Tocris Bioscience™ Supplier No. 0336/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-selective mGlu antagonist

Non-selective group I/group II metabotropic glutamate receptor antagonist.

S-enantiomer and Sodium Salt also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146669-29-6
Molecular Formula C10H11NO4

Specifications

Quantity 10 mg
Formula Weight 209.2
Purity >99%
Molecular Weight (g/mol) 209.201
Chemical Name or Material (RS)-MCPG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.