CAS RN 146669-29-6
IUPAC Name:
4-[(1R)-1-azaniumyl-1-carboxylatoethyl]benzoate
Synonyms:
4-[(1R)-1-ammonio-1-carboxylatoethyl]benzoate
Molecular Weight (g/mol):
208.19
Molecular Formula:
C10H10NO4
InChi Key:
DNCAZYRLRMTVSF-SNVBAGLBSA-M
SMILES:
C[C@]([NH3+])(C([O-])=O)C1=CC=C(C=C1)C([O-])=O
1
–
2
of
2
results
1
Tocris Bioscience™ (RS)-MCPG
CAS: 146669-29-6 Molecular Formula: C10H11NO4
2
(RS)-MCPG, MedChemExpress
MedChemExpress (RS)-MCPG (alpha-MCPG) is a competitive and selective group I/group II metabotropic glutamate receptor (mGluR) antagonist. (RS)-MCPG blocks theta-burst stimulation (TBS)-induced shifts in both juvenile and neonatal rat hippocampal neurons.