missing translation for 'onlineSavingsMsg'
Learn More

RPL7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen RPL7
Concentration 0.45 mg/mL
Applications Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16823294
View Documents
Proteintech
14583-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16813294
View Documents
Proteintech
14583-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NIP7 contains 1PUA domain and belongs to the NIP7 family. It may play a role in 60S ribosomal subunit synthesis.
Specifications

Specifications

RPL7
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Rabbit
Mouse, Rat, Human
P05426, P14148, P18124
19989, 297755, 6129
RPL7 Fusion Protein Ag6104
Primary
-20°C
RPL7
0.45 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
60S ribosomal protein L7, humL7 1, ribosomal protein L7, RPL7
RPL7
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.