missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14583-1-AP-150UL
This item is not returnable.
View return policy
Description
NIP7 contains 1PUA domain and belongs to the NIP7 family. It may play a role in 60S ribosomal subunit synthesis.Specifications
| RPL7 | |
| Polyclonal | |
| Unconjugated | |
| RPL7 | |
| 60S ribosomal protein L7, humL7 1, ribosomal protein L7, RPL7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19989, 297755, 6129 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05426, P14148, P18124 | |
| RPL7 | |
| RPL7 Fusion Protein Ag6104 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction