missing translation for 'onlineSavingsMsg'
Learn More

RPL31 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen RPL31
Concentration 0.13 mg/mL
Applications Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16896144
View Documents
Proteintech
16497-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16886144
View Documents
Proteintech
16497-1-AP-150UL
150 μL
£349.00
150µL
Ver productos alternativos
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPL31 is a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L31E family of ribosomal proteins. It is located in the cytoplasm.
Especificaciones

Especificaciones

RPL31
Immunocytochemistry, Western Blot, Immunofluorescence
Unconjugated
Rabbit
Mouse, Rat, Human
P62899, P62900, P62902
114641, 6160, 64298
RPL31 Fusion Protein Ag9648
Primary
-20°C
RPL31
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
60S ribosomal protein L31, ribosomal protein L31, RPL31
RPL31
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documentos

Documentos

Certificados
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado