missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL31 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16497-1-AP-150UL
This item is not returnable.
View return policy
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPL31 is a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L31E family of ribosomal proteins. It is located in the cytoplasm.Specifications
| RPL31 | |
| Polyclonal | |
| Unconjugated | |
| RPL31 | |
| 60S ribosomal protein L31, ribosomal protein L31, RPL31 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114641, 6160, 64298 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62899, P62900, P62902 | |
| RPL31 | |
| RPL31 Fusion Protein Ag9648 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction