missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPB8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-53016
This item is not returnable.
View return policy
Description
RPB8 Polyclonal specifically detects RPB8 in Human samples. It is validated for Western Blot.
Specifications
| RPB8 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DNA-directed RNA polymerases I, II, and III subunit RPABC3, hsRPB8, II, and III 17.1 kDa polypeptide, II, and III subunit ABC3, polymerase (RNA) II (DNA directed) polypeptide H, RPB8 homolog | |
| Rabbit | |
| 17 kDa | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: RPABC3. | |
| Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| Purified |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P52434 | |
| POLR2H | |
| Synthetic peptides corresponding to POLR2H(polymerase (RNA) II (DNA directed) polypeptide H) The peptide sequence was selected from the N terminal of human POLR2H. Peptide sequence DLGDKFRLVIASTLYEDGTLDDGEYNPTDDRPSRADQFEYVMYGKVYRIE. | |
| Protein A purified | |
| RUO | |
| 5437 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction