missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPB8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | RPB8 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693022
|
Novus Biologicals
NBP2-93995-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674482
|
Novus Biologicals
NBP2-93995-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPB8 Polyclonal antibody specifically detects RPB8 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| RPB8 | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neurodegeneration, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 5437 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DNA-directed RNA polymerases I, II, and III subunit RPABC3, hsRPB8, II, and III 17.1 kDa polypeptide, II, and III subunit ABC3, polymerase (RNA) II (DNA directed) polypeptide H, RPB8 homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human POLR2H (NP_006223.2). MAGILFEDIFDVKDIDPEGKKFDRVSRLHCESESFKMDLILDVNIQIYPVDLGDKFRLVIASTLYEDGTLDDGEYNPTDDRPSRADQFEYVMYGKVYRIEGDETSTEAATRLSAYVSYGGLLMRLQGDANNLHGFEVDSRVYLLMKKLAF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title