missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPB8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93995-0.1ml
This item is not returnable.
View return policy
Description
RPB8 Polyclonal antibody specifically detects RPB8 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| RPB8 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| DNA-directed RNA polymerases I, II, and III subunit RPABC3, hsRPB8, II, and III 17.1 kDa polypeptide, II, and III subunit ABC3, polymerase (RNA) II (DNA directed) polypeptide H, RPB8 homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human POLR2H (NP_006223.2). MAGILFEDIFDVKDIDPEGKKFDRVSRLHCESESFKMDLILDVNIQIYPVDLGDKFRLVIASTLYEDGTLDDGEYNPTDDRPSRADQFEYVMYGKVYRIEGDETSTEAATRLSAYVSYGGLLMRLQGDANNLHGFEVDSRVYLLMKKLAF | |
| 0.1 mL | |
| Neurodegeneration, Neuroscience | |
| 5437 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction