missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Round Specimen Clamps
Ensure easy insertion and removal of round specimens with Epredia™ Round Specimen Clamps.
£140.00 - £835.00
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12083059
|
Epredia™
546180 |
Round Specimen Clamp, No Inserts |
£357.00
Each |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12003069
|
Epredia™
546200 |
Insert For Clamping Jaw, 15 to 21mm |
£148.00
Each |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12013069
|
Epredia™
546210 |
Insert For Clamping Jaw, 22 to 28mm |
£153.00
Each |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12023069
|
Epredia™
546270 |
Insert For Clamping Jaw, 9 to 14.5mm |
£148.00
Each |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12093059
|
Epredia™
546190 |
Insert For Clamping Jaw, 6 to 9mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12017219
|
Epredia™
715580 |
Round specimen clamp, clamping case only, All inserts (6 to 28mm) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||