Learn More
Gilac™ Round Dividing Dough Container
Description
- Round dividing dough container.
- Highly resistant to changes in temperature.
- Highly flexible and shock-resistant.
- Compatible with racking trolleys.
Specifications
Specifications
| Type | Storage Tote |
| Material | HDPE |
| Color | White |
| Shape | Round |
| Capacity (English) | 5 gal. |
| Capacity (Metric) | 19 L |