missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf Rotor R12A6 for Centrifuge CR22N

Product Code. 31945295 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
4 x 500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
31945295 4 x 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31945295 Supplier Eppendorf Supplier No. 5721221010
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fixed-angle rotor designed for large-volume harvesting/pelleting of cells, bacteria, and algae.

  • Fixed-angle rotor for harvesting of bacteria, cells or yeast for e.g. recombinant protein purification or cell compartment purification
  • Includes 4 x 500 polypropylene bottle
  • Max. speed: 22,500 x g (12,000 rpm)
  • Max. capacity: 4 x 500 mL (Nominal capacity)
  • Made from high-quality aluminum
TRUSTED_SUSTAINABILITY

Specifications

Material Aluminum
Type Fixed Angle Rotor
Capacity 4 x 500 mL
Max. RCF 22,500 x g
Max. Speed 12,000 rpm
For Use With (Equipment) Centrifuge CR22N/CR21N
Biocontainment No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.