Learn More
Eppendorf Rotor R10H for Centrifuge CR30NX
Product Code. 31945081 Shop All Eppendorf Products
Change view
Click to view available options
Max. RCF:
13,000 x g
Unit Size:
Each
Product Code. Max. RCF Unit Size
31945081 13,000 x g Each
1 options
Product Code. 31945081 Supplier Eppendorf Supplier No. 5721302700
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specialized horizontal rotor compatible with the CR30NX centrifuge.
  • Horizontal rotor R10H
  • For plates
  • For centrifuge CR30NX
  • Max. speed: 13,000 x g (10,000 rpm)
  • For plate centrifugation for e.g. PCR or cell-free protein synthesis with up to 12 x 96 MTP
  • Made from high-quality aluminum
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Type Horizontal Rotor
Max. RCF 13,000 x g
Max. Speed 10,000 rpm
For Use With (Equipment) Centrifuge CR30NX
Biocontainment No