missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf Rotor R10C2 for Centrifuge CR30NX

Product Code. 31945189 Shop All Eppendorf Products
Change view
Click to view available options
Max. RCF:
14,290 x g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Max. RCF Unit Size
31945189 14,290 x g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31945189 Supplier Eppendorf Supplier No. 5721302601
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Continuous-flow rotor compatible with the CR30NX centrifuge.

  • Continuous flow rotor R10C2
  • Excludes top unit
  • For centrifuge CR30NX
  • Max. capacity: 3,200 mL (nominal capacity)
  • Max. speed: 14,290 x g (10,000 rpm)
  • For continuous flow harvesting of bacteria, cells or yeast for e.g. recombinant protein or antibody purification in very large volumes of less than 10 L
TRUSTED_SUSTAINABILITY

Specifications

Material Aluminum
Type Continuous Flow Rotor
Capacity 3,200 mL
Max. RCF 14,290 x g
Max. Speed 10,000 rpm
For Use With (Equipment) Centrifuge CR30NX
Biocontainment No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.