missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RosetteSep™ Human Total Lymphocyte Enrichment Cocktail
Description
- The RosetteSep™ Human Total Lymphocyte Enrichment Cocktail is designed to enrich lymphocytes from whole blood by negative selection
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD16, CD36, CD66b and glycophorin A on red blood cells (RBCs)
- When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
- The purified lymphocytes are present as a highly enriched population at the interface between the plasma and the buoyant density medium
Specifications
Specifications
| Cell Type | Lymphocytes |
| For Use With (Application) | Cell Isolation |
| Quantity | 200 mL whole blood |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?