missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktail
Description
- The RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktail is designed to isolate progenitor cells from cord blood and whole blood by negative selection
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD3, CD14, CD16, CD19, CD24, CD56, CD61, CD66b and glycophorin A on red blood cells (RBCs)
- When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
- The purified progenitor cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 200 mL cord blood |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?