missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR2 Antibody (6F2D10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£187.00 - £435.00
Specifications
| Antigen | ROR2 |
|---|---|
| Clone | 6F2D10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18122699
|
Novus Biologicals
NBP2-37263 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623626
|
Novus Biologicals
NBP2-37263-0.025ml |
0.025 mL |
£187.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROR2 Monoclonal specifically detects ROR2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| ROR2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2 | |
| ROR2 | |
| IgG1 | |
| Affinity Purified |
| 6F2D10 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Protein Kinase, Wnt Signaling Pathway | |
| PBS with 0.05% Sodium Azide | |
| 4920 | |
| Purified recombinant fragment of human ROR2 (AA: 59-155) expressed in E. Coli. | |
| Primary | |
| 104.8 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title