missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR2 Antibody (6F2D10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37263
This item is not returnable.
View return policy
Description
ROR2 Monoclonal specifically detects ROR2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| ROR2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2 | |
| Mouse | |
| 104.8 kDa | |
| 0.1 mL | |
| Protein Kinase, Wnt Signaling Pathway | |
| 4920 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 6F2D10 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ROR2 | |
| Purified recombinant fragment of human ROR2 (AA: 59-155) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction